EMAIL THIS PAGE TO A FRIEND
The European journal of contraception & reproductive health care : the official journal of the European Society of Contraception
Gestagens, danazol and antiprogestogen in emergency contraception.
PMID 9678102
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D8399
Danazol, ≥98%
C22H27NO2