EMAIL THIS PAGE TO A FRIEND
Journal of inherited metabolic disease
Phenotypic characterization of mice with targeted disruption of glycosylasparaginase gene: a mouse model for aspartylglycosaminuria.
PMID 9686358
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A6681
2-Acetamido-1-N-(β-L-aspartyl)-2-deoxy-β-D-glucopyranosylamine, ≥98%
C12H21N3O8