EMAIL THIS PAGE TO A FRIEND
Nucleosides & nucleotides
Some 6-aza-5-substituted-2'-deoxyuridines show potent and selective inhibition of herpes simplex virus type 1 thymidine kinase.
PMID 9708347
Abstract
The synthesis and X-ray crystal structures of a series of 5-substituted-6-aza-2'-deoxyuridines is reported. These nucleoside analogues inhibit the phosphorylation of thymidine by HSV-1 TK but have no effect on the corresponding human enzyme. Detailed examination of one analogue proves it to be a competitive inhibitor of thymidine with a Ki of 0.34 microM and is a very poor substrate. The analogues are not substrates for the enzyme and also do not inhibit the degradation of thymidine by thymidine phosphorylase. Molecular modelling showed that the inhibitors fit well in the active site of HSV-1 TK, provided the conformation of the sugar moiety is the same for thymidine in the complex.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A1882
6-Azauridine
C8H11N3O6