EMAIL THIS PAGE TO A FRIEND
Biochemistry
Spectroscopic characterization of intermediates in the urate oxidase reaction.
PMID 9709003
Abstract
The oxidation of urate catalyzed by soybean urate oxidase was studied under single-turnover conditions using stopped-flow absorbance and fluorescence spectrophotometry. Two discrete enzyme-bound intermediates were observed; the first intermediate to form had an absorbance maximum at 295 nm and was assigned to a urate dianion species; the second intermediate had an absorbance maximum at 298 nm and is believed to be urate hydroperoxide. These data are consistent with a catalytic mechanism that involves formation of urate hydroperoxide from O2 and the urate dianion, collapse of the peroxide to form dehydrourate, and hydration of dehydrourate to form the observed product, 5-hydroxyisourate. The rate of formation of the first intermediate was too fast to measure accurately at 20 degreesC; the second intermediate formed with a rate constant of 32 s-1 and decayed with a rate constant of 6.6 s-1. The product of the reaction, 5-hydroxyisourate, is fluorescent, and its release from the active site occurred with a rate constant of 31 s-1.