EMAIL THIS PAGE TO A FRIEND
Advances in carbohydrate chemistry and biochemistry
Applications of tin-containing intermediates to carbohydrate chemistry.
PMID 9710969
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
14509
Tin, ≥99%, powder
Sn
265640
Tin, powder, <150 μm, 99.5% trace metals basis
Sn
243434
Tin, granular, 0.425-2.0 mm particle size, ≥99.5%, ACS reagent
Sn
576883
Tin, nanopowder, <150 nm particle size (SEM), ≥99% trace metals basis
Sn
520373
Tin, powder, 10 μm, 99% trace metals basis
Sn
204692
Tin, shot, 99.999% trace metals basis
Sn
217697
Tin, 99.8%, shot, 3 mm
Sn
265632
Tin, powder, <45 μm particle size, 99.8% trace metals basis
Sn
265756
Tin, foil, thickness 0.5 mm, 99.998% trace metals basis
Sn
356948
Tin, foil, thickness 0.127 mm, 99.9%
Sn
755702
Tin, powder, -100 mesh, 99.99% trace metals basis
Sn
356956
Tin, wire, diam. 0.5 mm, 99.999% trace metals basis
Sn
GF96022852
Tin, foil, 2m coil, thickness 0.125mm, as rolled, 98.8%
Sn
GF53258182
Tin, foil, 25x25mm, thickness 1.0mm, as rolled, 99.99+%
Sn
GF48212669
Tin, foil, 100x100mm, thickness 0.125mm, as rolled, 98.8%
Sn
GF05063228
Tin, foil, 100x100mm, thickness 0.125mm, as rolled, 99.99+%
Sn
GF31638089
Tin, foil, 100x100mm, thickness 0.15mm, as rolled, 98.8%
Sn
GF21766068
Tin, foil, 100x100mm, thickness 0.15mm, as rolled, 99.99+%
Sn
GF04538617
Tin, foil, 100x100mm, thickness 0.1mm, as rolled, 98.8%
Sn
GF54272336
Tin, foil, 100x100mm, thickness 0.1mm, as rolled, 99.95%
Sn
GF79890889
Tin, foil, 100x100mm, thickness 0.20mm, as rolled, 98.8%
Sn
GF25742943
Tin, foil, 100x100mm, thickness 0.25mm, as rolled, 98.8%
Sn
GF11564535
Tin, foil, 100x100mm, thickness 0.25mm, as rolled, 99.99+%
Sn
GF45828827
Tin, foil, 100x100mm, thickness 0.5mm, as rolled, 98.8%
Sn
GF93825771
Tin, foil, 100x100mm, thickness 0.5mm, as rolled, 99.99+%
Sn
GF10675470
Tin, foil, 100x100mm, thickness 0.65mm, as rolled, 99.99+%
Sn
GF08231701
Tin, foil, 100x100mm, thickness 0.75mm, as rolled, 99.99+%
Sn
GF39022815
Tin, foil, 100x100mm, thickness 1.0mm, as rolled, 98.8%
Sn
GF59159223
Tin, foil, 100x100mm, thickness 1.0mm, as rolled, 99.99+%
Sn
GF98532041
Tin, foil, 100x100mm, thickness 1.0mm, as rolled, 99.999%
Sn