EMAIL THIS PAGE TO A FRIEND
The Journal of Biological Chemistry
A protein kinase C-, Ras-, and RSK2-dependent signal transduction pathway activates the cAMP-responsive element-binding protein transcription factor following T cell receptor engagement
PMID 9712919