EMAIL THIS PAGE TO A FRIEND
Gene
Isolation and characterization of cDNAs encoding PDE5A, a human cGMP-binding, cGMP-specific 3',5'-cyclic nucleotide phosphodiesterase.
PMID 9714779
Abstract
Human cGMP-binding, cGMP-specific 3',5'-cyclic nucleotide phosphodiesterase (PDE5A) cDNAs were isolated. A 3.1-kb composite DNA sequence assembled from overlapping cDNAs encodes an 875-amino-acid protein with a predicted molecular mass of 100012 Da (PDE5A1). Extracts prepared from yeast expressing human PDE5A1 hydrolyzed cGMP. This activity was inhibited by the selective PDE5 inhibitors zaprinast and DMPPO. PDE5A mRNA is expressed in aortic smooth muscle cells, heart, placenta, skeletal muscle and pancreas and, to a much lesser extent, in brain, liver and lung. A 5'-splice variant, PDE5A2, encodes an 833-amino-acid protein with eight unique amino acids at the amino terminus. PDE5A maps to chromosome 4q 25-27.