EMAIL THIS PAGE TO A FRIEND
Cell growth & differentiation : the molecular biology journal of the American Association for Cancer Research
Toward an understanding of the functional complexity of the E2F and retinoblastoma families.
PMID 9716176