EMAIL THIS PAGE TO A FRIEND
Journal of photochemistry and photobiology. B, Biology
Relative contributions of apoptosis and random necrosis in tumour response to photodynamic therapy: effect of the chemical structure of Zn(II)-phthalocyanines.
PMID 9718718
Abstract
Zn(II)-phthalocyanines (ZnPc) and its octapentyl (ZnOPPc) and octadecyl (ZnODPc) derivatives have been intravenously injected at a dose of 1.46 mumol/kg into female Balb/c mice bearing an intramuscularly transplanted MS-2 fibrosarcoma. Pharmacokinetic studies show that in all cases the maximal concentration of phthalocyanine in the tumour is reached at 24 h post-injection: the efficiency and selectivity of tumour targeting slightly increase upon increasing the length of the alkyl substituents. Irradiation of the neoplastic lesion (620-700 nm light, 180 MW/cm2, 300 J/cm2) 24 h after photosensitizer administration induces a significant delay of tumour growth, which was largest (approximately 11 days) for ZnPc and smallest (approximately 3.5 days) for ZnODPc. Electron microscopy investigations of irradiated tumour specimens show that ZnPc causes an early direct damage of malignant cells, largely via processes leading to random necrotic pathways, although a limited contribution of apoptotic pathways is detected. The importance of this increased upon using ZnOPPc and especially ZnODPc as the photosensitizers, possibly due to a different partitioning in different compartments of cell membranes.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
383813
Zinc 1,4,8,11,15,18,22,25-octabutoxy-29H,31H-phthalocyanine
C64H80N8O8Zn