EMAIL THIS PAGE TO A FRIEND
International journal of cancer
RARalpha antagonist Ro 41-5253 inhibits proliferation and induces apoptosis in breast-cancer cell lines.
PMID 9724098
Abstract
Ro 41-5253 is a RARalpha-selective antagonist that binds RARalpha but does not induce transcriptional activation and does not influence RAR/RXR heterodimerization and DNA binding. This retinoid inhibits proliferation and induces apoptosis in MCF-7 and ZR-75.1 estrogen-receptor-positive breast-carcinoma cells in a dose-dependent way. The anti-proliferative effect is more evident in ZR-75.1 cells than in MCF-7 cells and is probably mediated by anti-AP1 activity, a mechanism known to be implied in the action of several retinoids. In the induction of apoptosis also ZR-75.1 cells are more sensitive to treatment with Ro 41-5253 than MCF-7 cells. In ZR-75.1 cells an apoptotic/hypodiploid DNA peak is already evident after 2 days of incubation, whereas in MCF-7 cells it appears only after 4 days. The highest percentage of apoptotic cells, for both cell lines, is reached after 6 days of treatment. The apoptosis pathway is p53-independent and bcl-2 downregulation seems to be correlated with an increase in TGF-beta1 protein. The MDA-MB-231 estrogen-receptor-negative cell line is poorly responsive to Ro 41-5253 treatment, both in terms of proliferation inhibition and apoptosis induction. Ro 41-5253 has proliferation-inhibiting and apoptosis-inducing properties that are not mediated by transcriptional activation from retinoic-acid response elements. This retinoid antagonist seems to be a compound that exerts an anti-tumor activity but does not induce the toxic side effects of retinoids and might, therefore, be considered as a candidate for cancer therapy.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SML0573 Ro 41-5253, ≥98% (HPLC)
C28H36O5S