EMAIL THIS PAGE TO A FRIEND
Bulletin of environmental contamination and toxicology
Development of a 7-day Daphnia magna growth test using image analysis.
PMID 9724359
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
112151
2,4-Dichloroaniline, 99%
C6H5Cl2N
35829
2,4-Dichloroaniline, PESTANAL®, analytical standard
C6H5Cl2N