EMAIL THIS PAGE TO A FRIEND
Burns : journal of the International Society for Burn Injuries
Hypopigmentation along subcutaneous veins following intrakeloid triamcinolone injection: a case report and review of literature.
PMID 9725695
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
287334
Triamcinolone
C21H27FO6
T1899990
Triamcinolone, European Pharmacopoeia (EP) Reference Standard
C21H27FO6