EMAIL THIS PAGE TO A FRIEND
Environmental health perspectives
Three-dimensional visualization of physiologically based kinetic model outputs.
PMID 9738209
Abstract
Outputs from a physiologically based toxicokinetic (PB-TK) model for fish were visualized by mapping time-series data for specific tissues onto a three-dimensional representation of a rainbow trout. The trout representation was generated in stepwise fashion: 1) cross-section images were obtained from an anesthetized fish using a magnetic resonance imaging system, 2) images were processed to classify tissue types and eliminate unnecessary detail. 3) processed images were imported to a visualization software package (Application Visualization System) to create a three-dimensional representation of the fish, encapsulating five volumes corresponding to the liver, kidney, muscle, gastrointestinal tract, and fat, Kinetic data for the disposition of pentachloroethane in trout were generated using a PB-TK model. Model outputs were mapped onto corresponding tissues volumes, representing chemical concentration as color intensity. The workstation software was then used to animate the images, illustration the accumulation of pentachloroethane in each tissue during a continuous branchial (gill) exposure.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
P2000
Pentachloroethane, 95%
C2HCl5