EMAIL THIS PAGE TO A FRIEND
Journal of natural products
Antipruritic dinaphthofuran-7,12-dione derivatives from the pericarp of Impatiens balsamina.
PMID 9748380
Abstract
Dinaphthofuran-7,12-dione derivatives named balsaminones A (1) and B (2) were isolated from the pericarp of Impatiens balsamina L. together with the known compound 2-methoxy-1,4-naphthoquinone (3). Their structures were elucidated by spectral techniques. These compounds have significant antipruritic activity.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
189162
2-Methoxy-1,4-naphthoquinone, 98%
C11H8O3