EMAIL THIS PAGE TO A FRIEND
Immunity
The structure of HLA-DM, the peptide exchange catalyst that loads antigen onto class II MHC molecules during antigen presentation.
PMID 9768757
Abstract
The three-dimensional structure of the soluble ecto-domain of HLA-DM has been determined to 2.5 A resolution by X-ray crystallography. HLA-DM has both peptide exchange activity and acts as a chaperone to peptide-free class II MHC molecules. As predicted, the structure is similar to that of classical class II MHC molecules except that the peptide-binding site is altered to an almost fully closed groove. An unusual cavity is found at the center of the region that binds peptides in class II MHC molecules, and a tryptophanrich lateral surface is identified that is a candidate both for binding to HLA-DR, to effect catalysis, and to HLA-DO, an inhibitor.