EMAIL THIS PAGE TO A FRIEND
Journal of AOAC International
Determination of diethanolamine and N-nitrosodiethanolamine in fatty acid diethanolamides.
PMID 9772735
Abstract
Diethanolamine (DEA) is a precursor of N-nitrosodiethanolamine (NDELA), an animal carcinogen. A gas chromatographic (GC) method was developed for determining DEA in fatty acid diethanolamides that are commonly used in cosmetic products. Methanolic solutions of the amides were analyzed by GC with flame ionization detection on either a wide-bore methyl silicone (Rtx-1) or 95% dimethyl--5% diphenyl polysiloxane (SPB-5) capillary column. Recovery of DEA from fatty acid dialkanolamides at fortification levels of 0.50, 1.00, and 5.00% ranged from 94 to 100%. In a survey of commercial fatty acid diethanolamides, DEA was found at levels ranging from 1.1 to 14.0%, and most were in good agreement with manufacturer's DEA specifications. Fatty acid diethanolamides also were anlayzed for NDELA by liquid chromatography interfaced to a thermal energy analyzer. Recovery of NDELA from fatty acid diethanolamides at fortification levels of 50, 100, and 200 ppb averaged 95%. No NDELA was found in any of the fatty acid diethanolamide samples analyzed.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
N7632
N-Nitrosodiethanolamine
C4H10N2O3
24046 SPB®-1 Capillary GC Column, L × I.D. 60 m × 0.32 mm, df 0.25 μm
24166 SPB®-5 Capillary GC Column, L × I.D. 30 m × 0.20 mm, df 0.20 μm
24032 SPB®-5 Capillary GC Column, L × I.D. 15 m × 0.25 mm, df 0.25 μm
24034 SPB®-5 Capillary GC Column, L × I.D. 30 m × 0.25 mm, df 0.25 μm
24036 SPB®-5 Capillary GC Column, L × I.D. 60 m × 0.25 mm, df 0.25 μm
24033 SPB®-5 Capillary GC Column, L × I.D. 15 m × 0.25 mm, df 1.00 μm
24035 SPB®-5 Capillary GC Column, L × I.D. 30 m × 0.25 mm, df 1.00 μm
24037 SPB®-5 Capillary GC Column, L × I.D. 60 m × 0.25 mm, df 1.00 μm
24101-U SPB®-5 Capillary GC Column, L × I.D. 15 m × 0.32 mm, df 0.25 μm
24048 SPB®-5 Capillary GC Column, L × I.D. 30 m × 0.32 mm, df 0.25 μm
24050 SPB®-5 Capillary GC Column, L × I.D. 60 m × 0.32 mm, df 0.25 μm
24359 SPB®-5 Capillary GC Column, L × I.D. 25 m × 0.32 mm, df 0.52 μm
24100-U SPB®-5 Capillary GC Column, L × I.D. 15 m × 0.32 mm, df 1.00 μm
24049 SPB®-5 Capillary GC Column, L × I.D. 30 m × 0.32 mm, df 1.00 μm
24051 SPB®-5 Capillary GC Column, L × I.D. 60 m × 0.32 mm, df 1.00 μm
23307-U SPB®-5 Capillary GC Column, L × I.D. 50 m × 0.32 mm, df 5.00 μm
25316 SPB®-5 Capillary GC Column, L × I.D. 15 m × 0.53 mm, df 0.50 μm
25317 SPB®-5 Capillary GC Column, L × I.D. 30 m × 0.53 mm, df 0.50 μm
25383 SPB®-5 Capillary GC Column, L × I.D. 60 m × 0.53 mm, df 0.50 μm
25420-U SPB®-5 Capillary GC Column, L × I.D. 30 m × 0.53 mm, df 1.00 μm
25304 SPB®-5 Capillary GC Column, L × I.D. 15 m × 0.53 mm, df 1.50 μm
25305-U SPB®-5 Capillary GC Column, L × I.D. 30 m × 0.53 mm, df 1.50 μm
25389 SPB®-5 Capillary GC Column, L × I.D. 60 m × 0.53 mm, df 1.50 μm
25342 SPB®-5 Capillary GC Column, L × I.D. 15 m × 0.53 mm, df 3.00 μm
25343 SPB®-5 Capillary GC Column, L × I.D. 30 m × 0.53 mm, df 3.00 μm
25350 SPB®-5 Capillary GC Column, L × I.D. 60 m × 0.53 mm, df 3.00 μm
25346 SPB®-5 Capillary GC Column, L × I.D. 15 m × 0.53 mm, df 5.00 μm
25347 SPB®-5 Capillary GC Column, L × I.D. 30 m × 0.53 mm, df 5.00 μm
25351 SPB®-5 Capillary GC Column, L × I.D. 60 m × 0.53 mm, df 5.00 μm