EMAIL THIS PAGE TO A FRIEND
Eksperimental'naia i klinicheskaia farmakologiia
[Antihypoxants: their status and outlook].
PMID 9783115
Abstract
The survey deals with the up-to-date information in the literature on the problem of pharmacological correction of pathological changes found in the organism in the hypoxic syndrome. The principal approaches to pharmacotherapy of this pathological condition are pointed out. Information on the up-to-date and effective antihypoxia agents applied in medical practice and those in the stage of development is given.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
334677
2-Imino-4-thiobiuret, 99%
C2H6N4S