EMAIL THIS PAGE TO A FRIEND
European journal of pharmacology
Antiplatelet effect of Z-335, a new orally active and long-lasting thromboxane receptor antagonist.
PMID 9788773
Abstract
We investigated the pharmacological characteristics of Z-335 ((+/-)-sodium[2-[4-(chlorophenylsulfonylaminomethyl)indan-5-yl]ace tate monohydrate), a new indan derivative. Z-335 inhibited the specific binding of [3H]SQ-29548 to human platelets and guinea pig platelet membranes. The IC50 values of Z-335 for human platelets and guinea pig platelet membranes were 29.9 +/- 3.1 nM with a slope of 1.09 +/- 0.05 and 32.5 +/- 1.7 nM with a slope of 1.07 +/- 0.02, respectively. Z-335 inhibited thromboxane A2 receptor-mediated human and guinea pig platelet aggregation in vitro and oral administration of this drug to guinea pigs inhibited U-46619- and collagen-induced platelet aggregation for 24 h. Z-335 dose-dependently prevented the occurrence of U-46619-induced pulmonary thromboembolism in mice and the protective effect of this drug (0.3 and 3 mg/kg, p.o.) lasted for 24 h. These results strongly suggest that Z-335 is a potent, orally active and long-lasting thromboxane A2 receptor antagonist, which may be useful as an antiplatelet drug.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D7441
Daltroban, >98% (HPLC)
C16H16ClNO4S