EMAIL THIS PAGE TO A FRIEND
The Journal of biological chemistry
Interaction between citrate synthase and malate dehydrogenase. Substrate channeling of oxaloacetate.
PMID 9792662
Abstract
The interactions between pig heart citrate synthase and mitochondrial malate dehydrogenase or cytosolic malate dehydrogenase were studied using the frontal analysis method of gel filtration and by precipitation in polyethylene glycol. This method showed that an interaction between citrate synthase and mitochondrial malate dehydrogenase occurred but no interaction between citrate synthase and cytosolic malate dehydrogenase. Channeling of oxaloacetate in the malate dehydrogenase and citrate synthase-coupled systems was tested using polyethylene glycol precipitates of citrate synthase and mitochondrial malate dehydrogenase, and citrate synthase and cytosolic malate dehydrogenase. The effectiveness of large amounts of aspartate aminotransferase and oxaloacetate decarboxylase, as competing enzymes for the intermediate oxaloacetate, was examined. Aspartate aminotransferase and oxaloacetate decarboxylase were less effective competitors for oxaloacetate when precipitated citrate synthase and mitochondrial malate dehydrogenase in polyethylene glycol was used at low ionic strength compared with free enzymes in the absence of polyethylene glycol or with a co-precipitate of citrate synthase and cytosolic malate dehydrogenase. Substrate channeling of oxaloacetate with citrate synthase-mitochondrial malate dehydrogenase precipitate was inefficient at high ionic strength. These effects could be explained through electrostatic interactions of mitochondrial but not cytosolic malate dehydrogenase with citrate synthase.