EMAIL THIS PAGE TO A FRIEND
Zentralblatt fur Veterinarmedizin. Reihe A
Flumethasone-induced insulin resistance in calves.
PMID 9793474
Abstract
The impact of one single therapeutic dose of flumethasone (0.5 mg/100 kg b.w.) on insulin sensitivity was studied in calves. Hyperinsulinemic, euglycemic clamp tests were performed before and after flumethasone treatment. At 24 h after treatment, insulin-dependent glucose utilization was reduced by 74% (P < 0.005). No significant changes occurred 72 h post-treatment.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
F9507
Flumethasone
C22H28F2O5