EMAIL THIS PAGE TO A FRIEND
Journal of pharmaceutical and biomedical analysis
Simultaneous determination of pteridines in multicomponent mixtures using derivative spectrophotometry and partial least-squares calibration.
PMID 9800651
Abstract
Simple binary mixtures composed of very similar pteridines (neopterin and pterin) have been resolved by derivative spectrophotometry. Detection limits of 0.30 microgram ml-1 and 0.12 microgram ml-1 for pterin and neopterin, respectively, have been calculated. Also, different mixtures of pteridines considered as disease markers, such as pterin, neopterin, xanthopterin and isoxanthopterin, have been determined by using a partial least-squares (PLS-2) model. Calibration set containing 0-7 micrograms ml-1 for each component was used. The resolution of several mixtures and single determination was tested in artificial samples.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
I7388
Isoxanthopterin, ≥97.5%, powder
C6H5N5O2