EMAIL THIS PAGE TO A FRIEND
Journal of pharmaceutical and biomedical analysis
Capillary gas chromatographic assay of camphor and m-cresol in dermatological creams.
PMID 9800662
Abstract
Camphor and m-cresol mixtures are used in antiseptic and anti-itching creams. No compendial method exists for these preparations. This paper reports a capillary gas chromatographic method using FID detection with 2,6-di-tert-butyl-4-methylphenol as internal standard on a 30 m x 0.32 mm Supelcowax -10 column (0.25 micron film) with helium as carrier gas. Ramped temperature programming was applied. The method allows simultaneous quantitation of camphor and m-cresol in the presence of o- and p-cresols, calamine and zinc oxide. Overall percent recoveries (+/- SD, n = 9) of camphor, o-, p- and m-cresol from spiked placebo creams, at a labeled amount of 10 (w/w)% were 96.9 +/- 0.6, 98.2 +/- 0.6, 99.2 +/- 0.5 and 101.0 +/- 0.9%, respectively, and at a labeled amount of 1% were 96.7 +/- 0.6, 97.8 +/- 0.9, 97.8 +/- 0.6, and 100.3 +/- 1.0%, respectively. The recovery studies were carried out at +/- 30% of the labeled amounts. Linear peak area or height ratios were obtained (r > 0.999) for camphor, o-, p- and m-cresol covering a concentration range of 10-200% of the labeled amount. Linearity (r > 0.999) was also obtained for m-cresol when the relative concentration of o- and p-cresol was varied from 5 to 100% of the m-cresol concentration. The resolution between the 'critical pair' of p- and m-cresol was > or = 1.1. The limit of quantitation was 23 pg for m-cresol and 9.3 pg for camphor using an injection split of 1:50. The repeatability (%RSD) for all compounds were < 2% for peak area and < 1.4% for peak height ratios. System suitability and robustness of the method were established. The method was successively applied to the assay of available commercial products and allows assay of camphor and the three cresol isomers.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
40251-U 3-Methylphenol, certified reference material, 5000 μg/mL in methanol
442391
3-Methylphenol, analytical standard
C7H8O
65996
m-Cresol, analytical standard
C7H8O
C85727
m-Cresol, 99%
C7H8O
W353000
m-Cresol, ≥98%, FG
C7H8O
25026-U
SUPELCOWAX® 10 Capillary GC Column, L × I.D. 10 m × 0.10 mm, df 0.10 μm
24343 SUPELCOWAX® 10 Capillary GC Column, L × I.D. 15 m × 0.10 mm, df 0.10 μm
24169 SUPELCOWAX® 10 Capillary GC Column, L × I.D. 30 m × 0.20 mm, df 0.20 μm
24170 SUPELCOWAX® 10 Capillary GC Column, L × I.D. 60 m × 0.20 mm, df 0.20 μm
24077 SUPELCOWAX® 10 Capillary GC Column, L × I.D. 15 m × 0.25 mm, df 0.25 μm
24079 SUPELCOWAX® 10 Capillary GC Column, L × I.D. 30 m × 0.25 mm, df 0.25 μm
24081 SUPELCOWAX® 10 Capillary GC Column, L × I.D. 60 m × 0.25 mm, df 0.25 μm
23308-U SUPELCOWAX® 10 Capillary GC Column, df 0.25 μm
24284 SUPELCOWAX® 10 Capillary GC Column, L × I.D. 30 m × 0.25 mm, df 0.50 μm
24078 SUPELCOWAX® 10 Capillary GC Column, L × I.D. 15 m × 0.32 mm, df 0.25 μm
24080-U SUPELCOWAX® 10 Capillary GC Column, L × I.D. 30 m × 0.32 mm, df 0.25 μm
24082 SUPELCOWAX® 10 Capillary GC Column, L × I.D. 60 m × 0.32 mm, df 0.25 μm
24084 SUPELCOWAX® 10 Capillary GC Column, L × I.D. 30 m × 0.32 mm, df 0.50 μm
24085-U SUPELCOWAX® 10 Capillary GC Column, L × I.D. 60 m × 0.32 mm, df 0.50 μm
24211 SUPELCOWAX® 10 Capillary GC Column, L × I.D. 30 m × 0.32 mm, df 1.00 μm
24212 SUPELCOWAX® 10 Capillary GC Column, L × I.D. 60 m × 0.32 mm, df 1.00 μm
25325 SUPELCOWAX® 10 Capillary GC Column, L × I.D. 30 m × 0.53 mm, df 0.50 μm
25385 SUPELCOWAX® 10 Capillary GC Column, L × I.D. 60 m × 0.53 mm, df 0.50 μm
25300-U SUPELCOWAX® 10 Capillary GC Column, L × I.D. 15 m × 0.53 mm, df 1.00 μm
25301-U SUPELCOWAX® 10 Capillary GC Column, L × I.D. 30 m × 0.53 mm, df 1.00 μm
25391 SUPELCOWAX® 10 Capillary GC Column, L × I.D. 60 m × 0.53 mm, df 1.00 μm
25375-U SUPELCOWAX® 10 Capillary GC Column, L × I.D. 30 m × 0.53 mm, df 2.00 μm
25376 SUPELCOWAX® 10 Capillary GC Column, L × I.D. 60 m × 0.53 mm, df 2.00 μm
23327-U SUPELCOWAX® 10 Capillary GC Column, L × I.D. 30 m × 0.75 mm, df 1.00 μm