EMAIL THIS PAGE TO A FRIEND
The Journal of Biological Chemistry
Convergence of progesterone and epidermal growth factor signaling in breast cancer. Potentiation of mitogen-activated protein kinase pathways
PMID 9813039
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
06-588
Anti-STAT5A/B Antibody, serum, clone, Upstate®