EMAIL THIS PAGE TO A FRIEND
Veterinary parasitology
The poultry red mite Dermanyssus gallinae (De Geer, 1778): current situation and future prospects for control.
PMID 9823064
Abstract
The current importance of Dermanyssus gallinae, mainly in egg layer, in Europe is pointed out. The limits of conventional control methods are underlined (development of mite resistance--future insecticide legislation and animal welfare legislation). Alternative control methods such as feeding deterrents or biological control, are reviewed in the context of poultry pest management.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
46426
S-Hydroprene, PESTANAL®, analytical standard
C17H30O2
35029
Triflumuron, PESTANAL®, analytical standard
C15H10ClF3N2O3