EMAIL THIS PAGE TO A FRIEND
Clinical and experimental allergy : journal of the British Society for Allergy and Clinical Immunology
Urinary mediators and asthma.
PMID 9824399
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
M7780
3-Methylhistamine dihydrochloride, analytical standard
C6H11N3 · 2HCl