EMAIL THIS PAGE TO A FRIEND
International journal of legal medicine
Identification of famprofazone ingestion.
PMID 9826089
Abstract
After a traffic accident a 32-year-old man was suspected of having previously taken an illegal drug. An immunochemical screening procedure revealed positive results for amphetamines in both urine and blood samples. The preliminary test was confirmed by GC/MS and both amphetamine and methamphetamine were found in both body fluids. However, the man denied any use of drugs but claimed to have taken four tablets of Gewodin. One of the ingredients, famprofazone, undergoes metabolic conversion to amphetamine and methamphetamine. Using GC/ MS the ingestion of famprofazone was verified by identification of the unchanged parent compound in the urine sample.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
F8639
Famprofazone, analytical standard
C24H31N3O