Science (New York, N.Y.)
Purification and cloning of a protein kinase that phosphorylates and activates the polo-like kinase Plx1.
PMID 9831560
The Xenopus polo-like kinase 1 (Plx1) is essential during mitosis for the activation of Cdc25C, for spindle assembly, and for cyclin B degradation. Polo-like kinases from various organisms are activated by phosphorylation by an unidentified protein kinase. A protein kinase, polo-like kinase kinase 1 or xPlkk1, that phosphorylates and activates Plx1 in vitro was purified to near homogeneity and cloned. Phosphopeptide mapping of Plx1 phosphorylated in vitro by recombinant xPlkk1 or in progesterone-treated oocytes indicates that xPlkk1 may activate Plx1 in vivo. The xPlkk1 protein itself was also activated by phosphorylation on serine and threonine residues, and the kinetics of activation of xPlkk1 in vivo closely paralleled the activation of Plx1. Moreover, microinjection of xPlkk1 into Xenopus oocytes accelerated the timing of activation of Plx1 and the transition from G2 to M phase of the cell cycle. These results define a protein kinase cascade that regulates several events of mitosis.