EMAIL THIS PAGE TO A FRIEND
Deutsche medizinische Wochenschrift (1946)
[Hyperkalemia after trimethoprim or pentamidine. Until now, a little noticed side effect of antimicrobial therapeutic measures in AIDS patients].
PMID 9835895