EMAIL THIS PAGE TO A FRIEND
Bioorganic & medicinal chemistry
Synthesis and biological evaluation of 2-hydroxy derivatives of digitoxigenin and 3-epidigitoxigenin.
PMID 9839018
Abstract
The four stereoisomers of the 2-hydroxy derivatives of digitoxigenin and 3-epidigitoxigenin have been synthesized, their structures established by NMR, and their binding affinity for the digitalis receptor on Na+, K(+)-ATPase evaluated. These derivatives showed lower affinities than the parent compounds. The hydrophilic hydroxy groups in the alpha position are more detrimental to the affinity than hydroxy groups in the beta position.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D9404
Digitoxigenin
C23H34O4
241814
Lithium tri-tert-butoxyaluminum hydride solution, 1.0 M in THF
C12H28AlLiO3