EMAIL THIS PAGE TO A FRIEND
Bulletin of environmental contamination and toxicology
Sulcotrione soil metabolism in summer corn crops.
PMID 9841729
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
46318
Sulcotrione, PESTANAL®, analytical standard
C14H13ClO5S