EMAIL THIS PAGE TO A FRIEND
Journal of the American Society of Nephrology : JASN
Decreased aquaporin-2 expression and apical plasma membrane delivery in kidney collecting ducts of polyuric hypercalcemic rats.
PMID 9848772
Abstract
Hypercalcemia is frequently associated with a urinary concentrating defect and overt polyuria. The molecular mechanisms underlying this defect are poorly understood. Dysregulation of aquaporin-2 (AQP2), the predominant vasopressin-regulated water channel, is known to be associated with a range of congenital and acquired water balance disorders including nephrogenic diabetes insipidus and states of water retention. This study examines the effect of hypercalcemia on the expression of AQP2 in rat kidney. Rats were treated orally for 7 d with dihydrotachysterol, which produced significant hypercalcemia with a 15 +/- 2% increase in plasma calcium concentration. Immunoblotting and densitometry of membrane fractions revealed a significant decrease in AQP2 expression in kidney inner medulla of hypercalcemic rats to 45.7 +/- 6.8% (n = 11) of control levels (100 +/- 12%, n = 9). A similar reduction in AQP2 expression was seen in cortex (36.9 +/- 4.2% of control levels, n = 6). Urine production increased in parallel, from 11.3 +/- 1.4 to a maximum of 25.3 +/- 1.9 ml/d (P < 0.01), whereas urine osmolality decreased from 2007 +/- 186 mosmol/kg x H2O to 925 +/- 103 mosmol/kg x H2O (P < 0.01). Immunocytochemistry confirmed a decrease in total AQP2 labeling of collecting duct principal cells from kidneys of hypercalcemic rats, and reduced apical labeling. Immunoelectron microscopy demonstrated a significant reduction in AQP2 labeling of the apical plasma membrane, consistent with the development of polyuria. In summary, the results strongly suggest that AQP2 downregulation and reduced apical plasma membrane delivery of AQP2 play important roles in the development of polyuria in association with hypercalcemia.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D9257
Dihydrotachysterol, analytical standard
C28H46O