EMAIL THIS PAGE TO A FRIEND
Journal of AOAC International
Gas chromatographic determination of azoxystrobin, fluazinam, kresoxim-methyl, mepanipyrim, and tetraconazole in grapes, must, and wine.
PMID 9850581
Abstract
Azoxystrobin, fluazinam, kresoxim-methyl, mepanipyrim, and tetraconazole were determined in grapes, must, and wine by a gas chromatographic method with nitrogen-phosphorus (NP) and mass spectrometric (MS) detectors. Pesticides were isolated from the matrixes by online microextraction with acetone-hexane (50 + 50, v/v). Because of the high selectivity of NP and MS detectors, no interferent peaks were present and no cleanup was necessary. Recoveries from fortified grapes, must, and wine ranged from 80 to 111%, with coefficients of variation ranging from 1 to 14%. Limits of determination were 0.05 mg/kg for kresoxim-methyl and 0.10 mg/kg for the other compounds.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
34095
Fluazinam, PESTANAL®, analytical standard
C13H4Cl2F6N4O4
33970
Mepanipyrim, PESTANAL®, analytical standard
C14H13N3