EMAIL THIS PAGE TO A FRIEND
Neuroreport
Selected pyrethroid insecticides stimulate glutamate uptake in brain synaptic vesicles.
PMID 9855310
Abstract
We aimed to ascertain whether pyrethroid insecticides could influence the vesicular transport of the excitatory amino acid glutamate. The incubation of rat cortical synaptic vesicles with resmethrin and permethrin, consistently stimulated both ATP-dependent and -independent uptake of [3H]glutamate, while not evoking depletion of its vesicular content. Both processes were counteracted by valinomycin, a dissipator of the transmembrane potential gradient (deltapsi(sv)). Meanwhile, the vesicular influx of 36Cl- anions was impaired by pyrethroid concentrations which did not affect the ATP-dependent uptake of [14C]methylamine, as a marker for the proton gradient (deltapH). Thus, the stimulation of glutamate transport appeared to involve mainly the deltapsi(sv). A self-attenuating effect of selected pyrethroids on putatively enhanced excitatory transmission in severe intoxication is suggested.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
45655
Resmethrin, PESTANAL®, analytical standard
C22H26O3