EMAIL THIS PAGE TO A FRIEND
Hunan yi ke da xue xue bao = Hunan yike daxue xuebao = Bulletin of Hunan Medical University
[Establishment of mutant strain from human lung cancer cell (A549) with characteristics of resistance to 8-AG and HGPRT deficiency].
PMID 9868049
Abstract
A human lung cancer cell line (A549) mutant with hypoxanthine guanine phosphoribosyltransferase (HGPRT) deficiency and resistance to 8-AG was established by treatment of the A549 cells with 3 micrograms.ml-1 N-methyl-N'-nitro-nitrosoguanidine (MNNG), and prescreened in 0.25% agarose DMEM semisolid medium containing 20 micrograms.ml-1 8-AG. The mutant cells are resistant to cytotoxic effect of 8-AG and sensitive to hypoxanthine-aminopterin thymidine (HAT) selective medium. The mutant cells can be used as a candidate parental cells to fuse with the somatic cells.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A5284
8-Azaguanine, Hybri-Max, γ-irradiated, powder, BioXtra, suitable for hybridoma
C4H4N6O