EMAIL THIS PAGE TO A FRIEND
Mayo Clinic proceedings
Antifungal agents. Part I. Amphotericin B preparations and flucytosine.
PMID 9868423
Abstract
Traditionally, amphotericin B has been the cornerstone of antifungal treatment. Toxicity, however, is a major dose-limiting factor of amphotericin B deoxycholate. Nevertheless, it continues to have a major role in the treatment of deep-seated mycotic infections. Recently, less nephrotic lipid formulations, including amphotericin B lipid complex, amphotericin B cholesteryl sulfate, and liposomal amphotericin B, have been introduced. The pharmacologic properties, main indications, recommended dosages, related costs, and adverse effects of these various preparations are summarized in this review. Orally administered flucytosine is useful in certain infections, particularly cryptococcal meningitis, but it should be used with caution in patients with renal insufficiency.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
F0175000
Flucytosine, European Pharmacopoeia (EP) Reference Standard
C4H4FN3O