EMAIL THIS PAGE TO A FRIEND
Bioorganic & medicinal chemistry letters
Inhibitors of acyl-CoA:cholesterol O-acyltransferase (ACAT) as hypocholesterolemic agents: synthesis and structure-activity relationships of novel series of sulfonamides, acylphosphonamides and acylphosphoramidates.
PMID 9871672
Abstract
Sulfoacetic acid, phosphoramidate, and phosphoramide analogs of the ACAT inhibitors, CI-999 and CI-1011 were synthesized. The structure-activity relationships of these compounds as ACAT inhibitors are described.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
242802
Sulfoacetic acid, technical grade
C2H4O5S