EMAIL THIS PAGE TO A FRIEND
Bioorganic & medicinal chemistry letters
A membrane-permeant, bioactivatable derivative of Ins(1,3,4)P3 and its effect on Cl(-)-secretion from T84 cells.
PMID 9873447
Abstract
The synthesis of rac-2,5,6-tri-O-butyryl-myo-inositol 1,3,4-trisphosphate hexakis(acetoxymethyl) ester [Bt3-Ins(1,3,4)P3/AM, 1], a membrane-permeant derivative of myo-inositol 1,3,4-trisphosphate [Ins(1,3,4)P3] is reported. 1 inhibited calcium-mediated chloride secretion of T84 cells, suggesting a regulatory link of Ins(1,3,4)P3 and the biosynthesis of the known inhibitor myo-inositol 3,4,5,6-tetrakisphosphate.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
I3389
D-myo-Inositol 1,3,4-tris-phosphate ammonium salt
C6H15O15P3 · 6H3N