EMAIL THIS PAGE TO A FRIEND
Bioorganic & medicinal chemistry letters
The cholesterol metabolite cholest-4-en-3-one and its 3-oxo derivatives suppress body weight gain, body fat accumulation and serum lipid concentration in mice.
PMID 9873500
Abstract
Based on the findings that cholest-4-en-3-one, an intestinal metabolite of cholesterol, has an anti-obesity effect on animals, the structure-effect relationship of its 3-oxo derivatives and related compounds were investigated. Cholesten-3-ones, which possesses an enone structure with a carbonyl group at C3, markedly inhibit body weight gain and body fat accumulation, as well as the levels of serum triglyceride and cholesterol in animals without any clinical abnormalities.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
188174
(+)-4-Cholesten-3-one, 98%
C27H44O