EMAIL THIS PAGE TO A FRIEND
Bioorganic & medicinal chemistry letters
Detection of an anhydride intermediate in the carboxypeptidase A catalyzed hydrolysis of a peptide substrate by solid state NMR spectroscopy and its mechanistic implication.
PMID 9873738
Abstract
We have detected an anhydride intermediate in the CPA catalyzed proteolytic reaction of Gly-Tyr. It appears that since the zinc-bound water molecule which is believed to attack the scissile amide carbonyl carbon in the hydrolysis reaction is excluded by the N-terminal amino group of Gly-Tyr, the carboxylate of Glu-270 becomes to attack the amide bond to generate the anhydride intermediate.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
G3502
Gly-Tyr
C11H14N2O4