EMAIL THIS PAGE TO A FRIEND
The European Journal of Neuroscience
Subcellular and subsynaptic distribution of the NR1 subunit of the NMDA receptor in the neostriatum and globus pallidus of the rat: co-localization at synapses with the GluR2/3 subunit of the AMPA receptor.
PMID 9875351