EMAIL THIS PAGE TO A FRIEND
Drug development and industrial pharmacy
The composition of NF-defined emulsifiers: sorbitan monolaurate, monopalmitate, monostearate, monooleate, polysorbate 20, polysorbate 40, polysorbate 60, and polysorbate 80.
PMID 9876559
Abstract
Using the analytical constants for sorbitan monolaurate, monopalmitate, monostearate, and monooleate given in the National Formulary (NF), calculations were carried out that indicated that these emulsifiers are esters of sorbitol mono- and dianhydrides. Contrary to the NF description, no significant amount of sorbitol ester was calculated to be present, in agreement with recent experimental findings. Further calculations were made using the NF-defined analytical constants of polysorbate 20, polysorbate 40, polysorbate 60, and polysorbate 80, which indicate that these emulsifiers are esters primarily of polyoxyethylated sorbitol monoanhydride (i.e., sorbitan), with lesser quantities of polyoxyethylated esters of sorbitol dianhydride. Since all hydroxyl groups of the polysorbates are primary, random distribution of acyl groups on the available hydroxyls can be assumed, and the likely composition of these emulsifiers can be calculated. The most abundant compounds appear to be polyoxyethylated sorbitan mono-, di-, and triesters. Although the polysorbates are stated to contain 20 moles of ethylene oxide per mole of ester, the oxyethylene contents stated in the Food Chemicals Codex reveal that somewhat more than 20 moles of ethylene oxide are combined.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
388920
Sorbitan monopalmitate
C22H42O6