Description
Molecular Formula
Add to Cart
P7412
Pirenzepine dihydrochloride, ≥98% (TLC), powder
C19H21N5O2 · 2HCl
T122
Telenzepine dihydrochloride hydrate, solid, ≥98% (HPLC)
C19H22N4O2S · 2HCl · xH2O