EMAIL THIS PAGE TO A FRIEND
Die Pharmazie
Antimutagenicity of propolis.
PMID 9879573
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
N17351
Nitroguanidine
CH4N4O2