EMAIL THIS PAGE TO A FRIEND
Molecular and cellular biochemistry
Metabolism of the dimethyl ester of [2,3-(13)C]succinic acid in rat hepatocytes.
PMID 9879664
Abstract
Hepatocytes prepared from overnight fasted rats were incubated for 120 min in the presence of the dimethyl ester of [2,3-(13)C]succinic acid (10 mM). The identification and quantification of 13C-enriched metabolites in the incubation medium were performed by a novel computational strategy for the deconvolution of NMR spectra with multiplet structures and constraints. The generation of 13C-labelled metabolites, including succinate, fumarate, malate, lactate, alanine, aspartate and glucose accounted for about half of the initial amount of the ester present in the incubation medium. A fair correlation was observed between the experimental abundance of each 13C-labelled glucose isotopomer and the corresponding values derived from a model for the metabolism of [2,3-(13)C]succinate. Newly formed glucose was more efficiently labelled in the carbon C5 than C2, as well as the carbon C6 than C1, supporting the concept that D-glyceraldehyde-3-phosphate may undergo enzyme-to-enzyme channelling between glyceraldehyde-3-phosphate dehydrogenase and phosphofructoaldolase.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
112755
DBE-4 dibasic ester, 98%
C6H10O4
W239607
Dimethyl succinate, 98%, FG
C6H10O4