EMAIL THIS PAGE TO A FRIEND
Glycoconjugate journal
Studies on the reductive amination of 3-deoxy-D-manno-octulosonic acid (Kdo).
PMID 9881760
Abstract
Reductive amination of 3-deoxy-D-manno-octulosonic acid (Kdo) with allylamine (AIIN) or 2-(4-aminophenyl)ethylamine (APEA) yields epimer pairs of 2-N-allylamino and 2-N-[2-(4-aminophenyl)ethylamino]-2,3-dideoxy-D-glycero-D-galacto- and -2,3-dideoxy-D-glycero-D-talo-octonic acid. The yields were 50-60% due to reduction of Kdo to the respective polyols as side reaction products. Mass spectrometric analyses proved the amination derivatives to be the expected glycamines. Nuclear magnetic resonance (NMR) studies were performed on 2-N-allylamino-2,3-dideoxyoctonic acid which represents the chain terminus of allylaminated oligosaccharides derived from bacterial lipopolysaccharides (LPS) after acid hydrolysis and reductive allylamination.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
123056
4-(2-Aminoethyl)aniline, 97%
C8H12N2