EMAIL THIS PAGE TO A FRIEND
The American journal of surgical pathology
Primary nodal marginal zone lymphomas of splenic and MALT type.
PMID 9888704
Abstract
The existence of primary nodal marginal zone lymphomas (MZL) is controversial, as is their relationship to putative extranodal counterparts. Most nodal lymphomas with monocytoid B cell/marginal zone differentiation exhibit the morphologic and immunophenotypical characteristics of extranodal MALT-lymphomas. Splenic marginal zone lymphoma (SMZL) is also of putative marginal zone derivation, but it differs immunophenotypically from MALT lymphoma. To clarify the relationship between nodal and extranodal MZLs and to investigate the possible existence of a nodal variant of SMZL, 36 MZL initially considered to be primary nodal neoplasms were examined. Other low-grade lymphomas with marginal zone differentiation were excluded (small lymphocytic lymphoma/chronic lymphocytic leukemia [SLL/CLL], follicular lymphoma, and mantle cell lymphoma). Six nodal MZLs showed morphologic and phenotypic characteristics similar to those of SMZL, whereas 30 tumors were more similar to MALT-type lymphomas. The six tumors with SMZL features showed a polymorphic infiltrate surrounding residual germinal centers with absent or very attenuated mantle cuffs. These lymphomas were IgD positive (6/6) but cyclin D1 (0/5), CD5 (0/6), and CD23 (0/6) negative. Five of these patients came for treatment in stage I or II. No patient manifested splenomegaly, peripheral blood, and/or bone marrow infiltration either at diagnosis or during follow-up. Lymph nodes from 30 patients with MALT-type features showed a perisinusoidal and perivascular infiltration of monocytoid/centrocytoid cells and residual germinal centers with a relatively well-preserved mantle cuff. The neoplastic cells were negative for IgD (0/17), cyclin D1 (0/8), and CD5 (0/12). Seven of 16 (44%) patients with a detailed history and clinical follow-up had evidence of extranodal lymphoma. These observations suggest that most nodal B cell lymphomas with marginal zone differentiation are of the MALT type and that they are frequently associated with an extranodal component. In addition, a primary nodal counterpart of splenic MZL also exists, and may occur in the absence of splenomegaly.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
268A-1
IgD Rabbit Polyclonal Antibody