EMAIL THIS PAGE TO A FRIEND
Molecular genetics and metabolism
Isolated isobutyryl-CoA dehydrogenase deficiency: an unrecognized defect in human valine metabolism.
PMID 9889013
Abstract
A 2-year-old female was well until 12 months of age when she was found to be anemic and had dilated cardiomyopathy. Total plasma carnitine was 6 microM and acylcarnitine analysis while receiving carnitine supplement revealed an increase in the four-carbon species. Urine organic acids were normal. In vitro analysis of the mitochondrial pathways for beta oxidation, and leucine, valine, and isoleucine metabolism was performed in fibroblasts using stable isotope-labeled precursors to these pathways followed by acylcarnitine analysis by tandem mass spectrometry. 16-2H3-palmitate was metabolized normally down to the level of butyryl-CoA thus excluding SCAD deficiency. 13C6-leucine and 13C6-isoleucine were also metabolized normally. 13C5-valine incubation revealed a significant increase in 13C4-isobutyrylcarnitine without any incorporation into propionylcarnitine as is observed normally. These same precursors were also evaluated in fibroblasts with proven ETF-QO deficiency in which acyl-CoA dehydrogenase deficiencies in each of these pathways was clearly identified. These results indicate that in the human, there is an isobutyryl-CoA dehydrogenase which exists as a separate enzyme serving only the valine pathway in addition to the 2-methyl branched-chain dehydrogenase which serves both the valine and the isoleucine pathways in both rat and human.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
50405
2-Methylbutyryl-L-carnitine, ≥97.0% (HPLC)
C12H23NO4
51085
Isobutyryl-L-carnitine, ≥97.0% (TLC)
C11H21NO4