EMAIL THIS PAGE TO A FRIEND
Contributions to nephrology
Quantitative and qualitative assessment and clinical meaning of molecules removed with BK membranes.
PMID 9895437
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
F20505
2-Furoic acid, 98%
C5H4O3