EMAIL THIS PAGE TO A FRIEND
Fortschritte der Neurologie-Psychiatrie
[Opipramol (Insidon) in the treatment of somatoform disorders].
PMID 9922924
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
O5889
Opipramol dihydrochloride, ≥98% (HPLC), solid
C23H29N3O · 2HCl