EMAIL THIS PAGE TO A FRIEND
Annals of the New York Academy of Sciences
Parallel evaluation of 5-HT1A receptor localization and functionality: autoradiographic studies with [35S]-GTP gamma S and the novel, selective radioligand, [3H]-S 15535.
PMID 9928280
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
S5321
S15535, ≥98% (HPLC), solid
C21H24N2O2